Anti-Equine Serum Albumin Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
028988
USBI028988EA 505 EUR
USBI028988
Anti-Equine Serum Albumin Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: Equine Serum Albumin (Equine Serum Albumin)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR