Anti-FAM65C Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
035473-HRP
USBI035473-HRPEA 884 EUR
USBI035473-HRP
Anti-FAM65C Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: FAM65C (family with sequence similarity 65, member C)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol FAM65C
Environmentally Preferable
Antigen name Family with sequence similarity 65, member C
Antibody type Primary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitope C-terminal
Amino acid number 699 to 729
Form liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR