Anti-IGLL5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
036922-HRP
USBI036922-HRPEA 884 EUR
USBI036922-HRP
Anti-IGLL5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: IGLL5 (immunoglobulin lambda-like polypeptide 5)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol IGLL5
Environmentally Preferable
Antigen name immunoglobulin lambda-like polypeptide 5
Antibody type Primary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitope C-terminal
Amino acid number 166 to 193
Form liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR