Anti-GMPR Mouse Polyclonal Antibody

Supplier: US Biological
127402
USBI127402EA 655 EUR
USBI127402
Anti-GMPR Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GMPR (guanosine monophosphate reductase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol GMPR
Environmentally Preferable
Antigen name Guanosine monophosphate reductase
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Mouse
Isotype IgG
Western blot Yes
Amino acid number 1 to 345
Sequence MPRIDADLKLDFKDVLLRPKRSSLKSRAEVDLERTFTFRNSKQTYSGIPIIVANMDTVGTFEMAAVMSQHSMFTAIHKHYSLDDWKLFATNHPECLQNVAVSSGSGQNDLEKMTSILEAVPQVKFICLDVANGYSEHFVEFVKLVRAKFPEHTIMAGNVVTGEMVEELILSGADIIKVGVGPGSVCTTRTKTGVGYPQLSAVIECADSAHGLKGHIISDGGCTCPGDVAKAFGAGADFVMLGGMFSGHTECAGEVIERNGRKLKLFYGMSSDTAMNKHAGGVAEYRASEGKTVEVPYKGDVENTILDILGGLRSTCTYVGAAKLKELSRRATFIRVTQQHNTVFS
Form liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR