Anti-GPR176 Mouse Polyclonal Antibody

Supplier: US Biological
127497
USBI127497EA 655 EUR
USBI127497
Anti-GPR176 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GPR176 (G protein-coupled receptor 176)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR