Anti-ADA Rabbit Polyclonal Antibody

Supplier: US Biological
A0870-20M
USBIA0870-20MEA 632 EUR
USBIA0870-20M
Anti-ADA Rabbit Polyclonal Antibody
Antibodies
  • Recommended dilution: ELISA: 1:1000; Immunochemistry: 10 μg/ml

Type: Primary
Antigen: ADA (adenosine deaminase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR