Anti-ADM2 Rabbit Polyclonal Antibody (Carboxyfluorescein)

Supplier: US Biological
A0909-42L2
USBIA0909-42L2EA 1160 EUR
USBIA0909-42L2
Anti-ADM2 Rabbit Polyclonal Antibody (Carboxyfluorescein)
Antibodies
Type: Primary
Antigen: ADM2 (adrenomedullin 2)
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR