Anti-Serum Albumin Sheep Polyclonal Antibody

Supplier: US Biological
A1327-02
USBIA1327-02EA 761 EUR
USBIA1327-02
Anti-Serum Albumin Sheep Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:50000-1:200000

Type: Primary
Antigen: Serum Albumin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR