Anti-Arginine, symmetric, dimethyl Rabbit Polyclonal Antibody

Supplier: US Biological
A3550-07C
USBIA3550-07CEA 863 EUR
USBIA3550-07C
Anti-Arginine, symmetric, dimethyl Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: A 1:500-1:1000

Type: Primary
Antigen: Arginine, symmetric, dimethyl
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR