Anti-CRK Rabbit Polyclonal Antibody (FITC (Fluorescein))

Supplier: US Biological
C7924-10E-FITC
USBIC7924-10E-FITCEA 884 EUR
USBIC7924-10E-FITC
Anti-CRK Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antibodies
Recommended dilution: FLISA: 1:1000 Dot Blot: 1:500
Do not freeze!

Type: Primary
Antigen: CRK (v-crk avian sarcoma virus CT10 oncogene homolog)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR