Anti-Cyclophilin H Chicken Polyclonal Antibody

Supplier: US Biological
C8855-86
USBIC8855-86EA 474 EUR
USBIC8855-86
Anti-Cyclophilin H Chicken Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:250; Immunochemistry: 1:100

Type: Primary
Antigen: Cyclophilin H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR