Anti-Immunoglobulin-Like Transcript 2 Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 7H15]

Supplier: US Biological
I7502-70A
USBII7502-70AEA 640 EUR
USBII7502-70A
Anti-Immunoglobulin-Like Transcript 2 Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 7H15]
Antibodies
Type: Primary
Antigen: Immunoglobulin-Like Transcript 2
Clonality: monoclonal
Clone: 7H15
Conjugation: Carboxyfluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name Immunoglobulin-Like Transcript 2
Antibody type Primary
Clonality Monoclonal
Conjugation FAM (Carboxyfluorescein)
Clone 7H15
Reactivity Human
Host Mouse
Isotype IgG1
Flow cytometry Yes
Amino acid number 24 to 458
Form liquid
Purification purified by Protein G chromatography
Storage buffer PBS, 0,5% BSA, 0,1% sodium azide.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR