Anti-Interleukin 17 Receptor Goat Polyclonal Antibody (Carboxyfluorescein)

Supplier: US Biological
I8439-35F
USBII8439-35FEA 524 EUR
USBII8439-35F
Anti-Interleukin 17 Receptor Goat Polyclonal Antibody (Carboxyfluorescein)
Antibodies
Type: Primary
Antigen: Interleukin 17 Receptor
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR