Anti-IVL Mouse Monoclonal Antibody [clone: 10K258]

Supplier: US Biological
I8447-27F
USBII8447-27FEA 539 EUR
USBII8447-27F
Anti-IVL Mouse Monoclonal Antibody [clone: 10K258]
Antibodies
Recommended dilution: Western Blot: 2?g/ml; Immunochemistry: 0.4-1 ?g/ml

Type: Primary
Antigen: IVL (involucrin)
Clonality: monoclonal
Clone: 10K258
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR