Anti-Melanocortin Receptor 4 Rabbit Polyclonal Antibody (Carboxyfluorescein)

Supplier: US Biological
M2868-39D2
USBIM2868-39D2EA 836 EUR
USBIM2868-39D2
Anti-Melanocortin Receptor 4 Rabbit Polyclonal Antibody (Carboxyfluorescein)
Antibodies
Type: Primary
Antigen: Melanocortin Receptor 4
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR