Anti-Netrin G1 Ligand Precursor Sheep Polyclonal Antibody (Biotin)

Supplier: US Biological
N1193-10M
USBIN1193-10MEA 910 EUR
USBIN1193-10M
Anti-Netrin G1 Ligand Precursor Sheep Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: Netrin G1 Ligand Precursor
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR