Anti-Neurofilament Light Mouse Monoclonal Antibody [clone: 9G400]

Supplier: US Biological
N2160-01B1
USBIN2160-01B1EA 542 EUR
USBIN2160-01B1
Anti-Neurofilament Light Mouse Monoclonal Antibody [clone: 9G400]
Antibodies
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 2.0-4.0 ?g/ml

Type: Primary
Antigen: Neurofilament Light
Clonality: monoclonal
Clone: 9G400
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR