(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) ≥99% for molecular biology

Supplier: US Biological

O7000
USBIO7000EA 101 EUR
USBIO7000
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) ≥99% for molecular biology
(2-Nitrophenyl) β-D-galactopyranoside

A colorimetric substrate for beta-D-glucuronidase used in the identification of E. coli and Salmonella.

Formula: C₁₂H₁₅NO₈
Storage Temperature: Refrigerator
MDL Number: MFCD00063255
CAS Number: 369-07-3

Order Now

Specification Test Results

Appearance White to slight yellow powder
Purity ≥99%
Identity (IR) Conforms to reference
Solubility (1%, H₂O) Colorless to slight yellow, clear, complete DMSO (slightly), water (slightly, sonicated)
Turbidity (1%, H₂O) ≤4 NTU
Water (KF) ≤0,5%
Specific rotation (C=1, H₂O) [a]D: −68,0 ± 3°
Free o-nitrophenol ≤0,03%
Bromine ≤2300 mg/kg
Aluminum ≤560 mg/kg

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR