Anti-PMP22 Rabbit Polyclonal Antibody

Supplier: US Biological
P4305-04B
USBIP4305-04BEA 703 EUR
USBIP4305-04B
Anti-PMP22 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: PMP22 (peripheral myelin protein 22)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol PMP22
Environmentally Preferable
Antigen name peripheral myelin Protein 22
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype
Epitope C-terminal
Form liquid
Purification purified by immunoaffinity chromatography
Concentration ~0,1 mg/ml
Storage buffer PBS, pH 7,4, 0,2% BSA, 0,05% sodium azide.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR