Anti-UCHL1 Rabbit Polyclonal Antibody

Supplier: US Biological
P9102-71K
USBIP9102-71KEA 667 EUR
USBIP9102-71K
Anti-UCHL1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol UCHL1
Environmentally Preferable
Antigen name Ubiquitin Carboxyl-Terminal Esterase L1
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitope N-terminal
Form liquid
Purification purified by Protein G chromatography
Storage buffer PBS, 0,09% sodium azide.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR