Anti-QRFP Receptor Rabbit Polyclonal Antibody (Carboxyfluorescein)

Supplier: US Biological
Q0575-55C
USBIQ0575-55CEA 1160 EUR
USBIQ0575-55C
Anti-QRFP Receptor Rabbit Polyclonal Antibody (Carboxyfluorescein)
Antibodies
Type: Primary
Antigen: QRFP Receptor
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR