Anti-Siglec 3 Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 7H35]

Supplier: US Biological
S1013-58A
USBIS1013-58AEA 483 EUR
USBIS1013-58A
Anti-Siglec 3 Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 7H35]
Antibodies
Type: Primary
Antigen: Siglec 3
Clonality: Monoclonal
Clone: 7H35
Conjugation: Carboxyfluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR