Anti-Splicing Factor, Arginine/Serine-Rich 3 Rabbit Polyclonal Antibody

Supplier: US Biological
S5554-54-100
USBIS5554-54-100EA 417 EUR
USBIS5554-54-100
Anti-Splicing Factor, Arginine/Serine-Rich 3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 50000

Type: Primary
Antigen: Splicing Factor, Arginine/Serine-Rich 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR