Anti-TIMP1 Mouse Monoclonal Antibody

Supplier: US Biological
T5581-02B
USBIT5581-02BEA 513 EUR
USBIT5581-02B
Anti-TIMP1 Mouse Monoclonal Antibody
Antibodies
Recommended dilution: Western Blot: 4?g/ml

Type: Primary
Antigen: TIMP1 (TIMP metallopeptidase inhibitor 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Monkey
Order Now

SPECIFICATIONS

Antigen symbol TIMP1
Antigen name TIMP metallopeptidase inhibitor 1
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Reactivity Monkey,Human
Host Mouse
Isotype IgG1 kappa
Western blot Yes
ELISA Yes
Form liquid
Purification purified by immunoaffinity chromatography
Concentration ~0,5 mg/ml
Storage buffer PBS, pH 7,2, 0,09% sodium azide.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR