Safety spectacles, uvex x-fit
Supplier: UVEX
9199.085
UVEX9199.085EA 20.5 EUR
UVEX9199.085
Safety spectacles, uvex x-fit
Safety Glasses
The model x-fit are classic safety spectacles with good coverage of the eyes and a high level of wearer acceptance.
  • Supravision coating technology
  • Metal-free
  • Translucent side arms
Weight: 23 g
Certifications: EN 166; EN 170