UVP crosslinker, CL-3000
Supplier: UVP ULTRA VIOLET PRODUCTS
849-95-0615-06 849-95-0615-02 849-95-0615-04
UVPA849-95-0615-06EA 2410 EUR
UVPA849-95-0615-06 UVPA849-95-0615-02 UVPA849-95-0615-04
UVP crosslinker, CL-3000
Lights UV Lights, Lamps and Bulbs
Ultraviolet light is considered an ancillary disinfectant to chemical disinfectants in the healthcare environment.
  • Perfect for delivering the recommended >1 J/cm² germicidal UV dose for surface disinfection
  • Crosslinking of DNA and RNA to nylon or nitrocellulose membranes
  • Microprocessor control provides precise UV dose control
  • Irradiation can be defined as energy (Joules/cm²) or time (seconds)
  • Bulbs: 8 W (6)
  • Weight: 6,8 kg
The high-precision CL-3000 crosslinker is capable of producing a cumulative dosage of 1 J/cm², this is considered well beyondMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable