Vacuum controller
Supplier: VACUUBRAND
20683180
VACU20683180EA 2710 EUR
VACU20683180
Vacuum controller
Pumps Vacuum Controllers
The VACUU.LAN local vacuum network connects several workstations to one vacuum pump. The various modules make it possible to connect almost all popular vacuum applications such as rotary evaporators, vacuum concentrators, gel dryers, drying ovens and filter systems. It can be integrated into new laboratory furniture at the time of assembly or subsequently installed in existing laboratories.
  • Individual connection configuration, vacuum connections to suit the requirements of the workstation
  • Integrated non-return valves to prevent cross-contamination, even
More Product Information