Solid phase extraction cartridges, Bond Elut™ PBA
Supplier: VARIAN
12113018 12105033 12102018 12102019 12102105 12102127
551-0344EA 445 EUR
551-0344 VARI12105033 VARI12102018 VARI12102019 VARI12102105 VARI12102127
Solid phase extraction cartridges, Bond Elut™ PBA
Chromatography Columns
PBA is a unique sorbent consisting of a phenylboronic acid covalently linked to a silica gel surface. Covalent processes result in a higher retention to the sorbent surface than other mechanisms, resulting in superior clean-up. The boronate group has a high specificity for cis-diol containing compounds such as catechols, nucleic acids, low molecular weight proteins and carbohydrates.
  • High specificity for cis-diol compounds
  • Very effective in separating DNA and RNA
Bond Elut™ PBA can also be used to retain oligomeric materials such as polyethyleneMore Product Information