PBA plates, Bond Elut
Supplier: VARIAN
7541201C
VARI7541201CEA 1130 EUR
VARI7541201C
PBA plates, Bond Elut
Microplates Solid Phase Extraction Plates (SPE)
Unique bond Elut PBA silica sorbent uses phenylboronic acid functionality to retain analytes via a reversible covalent bond, with high specificity and cleanliness.
  • Unique phenylboronic acid sorbent
  • High specificity for cis-diol compounds
  • Amenable for a broad range of biomolecule applications
Bond Elut PBA is a unique silica SPE sorbent containing a phenylboronic acid functionality that can retain analytes through a reversible covalent bond. This very strong covalent retention mechanism enables high specificity and cleanliness.
The boronate groupMore Product Information