GC Columns, J&W CP-Sil 43 CB
Supplier: VARIAN
CP7745
VARICP7745EA 904 EUR
VARICP7745
GC Columns, J&W CP-Sil 43 CB
Chromatography Columns
CP-Sil 43 CB separates aromatic from aliphatic hydrocarbons with moderate polarity for specific selectivity equivalent to OV-255.
  • 25% cyanopropyl/25% phenyl/50% dimethylpolysiloxane phase
  • Mid polarity
  • Solvent rinsable
Agilent JW CP-Sil 43 CB is a chemically bonded and cross-linked, moderately polar column.