High volume magnetic stirrers
Supplier: VELP SCIENTIFIC
F203A0511 F203A0513 F203A0510 F203A0512
VELPF203A0511EA 616 EUR
VELPF203A0511 VELPF203A0513 VELPF203A0510 VELPF203A0512
High volume magnetic stirrers
Stirrers Magnetic Stirrers
Magnetic stirrers that guarantee continuous operation at both low and high speeds ensuring safety and resistance to chemical agents.
  • The brushless motor provides continuous operation with high liquid volumes at any speed
  • Premium power and long life span
  • Constant stirring from 40 to 1500 rpm (MSL 8 - 25) or from 20 to 1100 (MSL 50) with SpeedServo technology
  • Gentle start and low speed to protect samples
  • The control panel is protected from liquid spills
  • Resistant stainless steel plate are
  • Designed for a long lifespan and guaranteed with 2 years warranty
More Product Information