Tongs, Chemware®
Supplier: Saint Gobain Life Sciences
D1069290
VERND1069290EA 91.5 EUR
VERND1069290
Tongs, Chemware®
Tongs
These stainless steel tongs are covered with an extra-heavy, non stick PTFE coating. For handling hot, cold, corrosive or sticky materials.
  • No loss of sample through sticking
  • Will not scratch or damage delicate surfaces
  • Contoured jaw enables handling of crucibles and other objects