Sani-Tech® STHT–C Tubings, Silicone, Platinum Cured
Supplier: Saint Gobain Life Sciences
STHT-C-187-2F STHT-C-187-2H STHT-C-250-3F STHT-C-250-3H STHT-C-312-4F STHT-C-375-2H STHT-C-375-4F STHT-C-375-4H HSTHT-C-1000-6
VERNSTHT-C-187-2FEA 5390 EUR
VERNSTHT-C-187-2F VERNSTHT-C-187-2H VERNSTHT-C-250-3F VERNSTHT-C-250-3H VERNSTHT-C-312-4F VERNSTHT-C-375-2H VERNSTHT-C-375-4F VERNSTHT-C-375-4H VERNHSTHT-C-1000-6
Sani-Tech® STHT–C Tubings, Silicone, Platinum Cured
Tubing Transfer Tubing
STHT®-C silicone tubing is an ultra-pure, biopharmaceutical-grade tubing which resists temperature extremes, ozone, corona, radiation, moisture, compression sets, weathering and chemical attack and imparts no taste or odours to fluids transported within it.
  • Unsurpassed surface smoothness
  • Ultra-flexible
  • Temperature resistance from –62 to 260 °C
Permanently laser marked with part number and lot code for total product traceability. Used in sterile fill and transfers, drug delivery, media processing, sanitary peristaltic pump tubing, laboratory use, cellMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable