Magnetic stirring bars, PTFE, cross-shaped
Supplier: VITLAB
316497
VITL316497EA 0 EUR
VITL316497
Magnetic stirring bars, PTFE, cross-shaped
Stirrers Magnetic Stirring Bars
These cross shaped magnetic stirrer bars have a magnetic core of Alnico 5 magnet encapsulated in PTFE. The angled shape gives rise to significant turbulence, and thus achieves effective mixing, even at low spin rates.
  • Cross shape promotes very stable centering