Water jet pumps, PP
Supplier: VITLAB
77094
VITL77094EA 0 EUR
VITL77094
Water jet pumps, PP
Pumps Vacuum Water Jet Pumps
This water jet vacuum pump for generation of a vacuum and to siphon off liquids and steam. It is chemical resistant, and the fluid path consists of PP, FKM and PTFE.
  • Integrated non-return valve increases safety
  • Simple operation and easy to clean
  • Detachable vacuum connection
Delivery information: Supplied with adapters that simplify connections, reducing adapters R ¹/₂", water connection sleeve nut R ³/₄", tubing connector (ext. Ø 10 - 12 mm), detachable hose nozzle (ext. Ø 6 - 9 mm) with screw cap (GL 14).