Screw caps
Supplier: VITLAB
83310 83311 83312 83313 83314 83315 83316 83317
VITL83310EA 26.6 EUR
VITL83310 VITL83311 VITL83312 VITL83313 VITL83314 VITL83315 VITL83316 VITL83317
Screw caps
Closures Bottle Closures
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.
Order Now
SPECIFICATIONS
Environmentally Preferable