Ammonia solution 10%, AnalaR NORMAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

190064VEA 60.2 EUR
190064V VWRC190065W 190063U
Ammonia solution 10%, AnalaR NORMAPUR®
Ammonia solution
Formula: NH₃
MW: 35.05 g/mol
Density: 0.957 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now

Specification Test Results

Assay Min. 10.0 %
Appearance Clear colourless liquid
Cl (Chloride) Max. 0.5 ppm
PO₄ (Phosphate) Max. 0.1 ppm
S (Sulphide) Max. 0.1 ppm
SO₄ (Sulphate) Max. 0.1 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 1 ppm
Mg (Magnesium) Max. 0.5 ppm
Mn (Manganese) Max. 0.1 ppm
Na (Sodium) Max. 1 ppm
Ni (Nickel) Max. 0.05 ppm
Pb (Lead) Max. 0.05 ppm
Zn (Zinc) Max. 0.2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR