Anti-CD7 Rabbit Monoclonal Antibody [clone: EP132]

Supplier: IMMUNOLOGIC
VWRKILM3007-C01EA 0 EUR
VWRKILM3007-C01 VWRKILM3007-C05 VWRKILM3007-C1
Anti-CD7 Rabbit Monoclonal Antibody [clone: EP132]
Antibodies
Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: EP132
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR