Cerium(IV) sulphate 0.05 mol/l (0.05 N) aqueous solution, AVS TITRINORM Volumetric

Supplier: VWR Collection
Warning

Synonyms: Cerium disulphate, Ceric sulphate, Cerium(IV) sulphate

VWRS31307.460EA 0 EUR
VWRS31307.460
Cerium(IV) sulphate 0.05 mol/l (0.05 N) aqueous solution, AVS TITRINORM Volumetric
Cerium (IV) sulphate
Formula: Ce(SO₄)₂
MW: 332.24 g/mol
Melting Pt: 195
MDL Number: MFCD00148852
CAS Number: 13590-82-4
EINECS: 237-029-5
UN: 3264
ADR: 8 III
Merck Index: 13,02002

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.0499 - 0.0501 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR