Fuchsin (acid), Gurr™ Microscopy_Staining

Supplier: VWR Collection

Synonyms: Rubin S, CN DcR-8, Acid Violet 19, Acid Fuchsin, Acid Fuchsin sodium salt, Fuchsin S, CI 42685, Acid Magenta, 3-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acid, Benzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)

VWRS34319.5000EA 0 EUR
VWRS34319.5000
Fuchsin (acid), Gurr™ Microscopy_Staining
Fuchsin (acid)
Formula: C₂₀H₁₇N₃Na₂O₉S₃
MW: 585.55 g/mol
Melting Pt: > 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00013286
CAS Number: 3244-88-0
EINECS: 221-816-5
Merck Index: 12,00109

Order Now

Specification Test Results

Dye content (titanometric) Min 60 %
Suitability for microscopy Passes test
Loss on drying (110°C) Max 10 %
Absorptivity (A 1%/1cm, Lambda max., in dilute HCl) 780 to 1310
Absorption max. Lambda (in dilute HCl) 540 to 549 nm
TLC test Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR