Sulphuric acid 84.5% 84,5% 84,5, VOLUSOL® ISO Volumetric

Supplier: VWR Collection
Danger

VWRS83958.330EA 0 EUR
VWRS83958.330
Sulphuric acid 84.5% 84,5% 84,5, VOLUSOL® ISO Volumetric
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR