VWR® Sample Sak™, Sample bags
Supplier: VWR Collection
11216-000
VWRU11216-000EA 180 EUR
VWRU11216-000
VWR® Sample Sak™, Sample bags
Bags Sample Bags
Ideal for collection and transport containers for powder and granular samples including grain, seeds, feeds, coffee, powdered soaps, and even damp soil.
  • For solid and semi-solid samples
  • Heat-sealed
  • Imprinted writing area
  • Lined with 1,25 mm polyethylene
  • Machine finished white Kraft paper with tin tie tabs
Constructed of 22,7 kg white Kraft paper. Imprinted on each bag is a personalised area for recording sample data and date.