VWR® Talon®, Clamps, Three Pronged
Supplier: VWR Collection
12621-236 21570-127 12621-238 21570-404 12621-240 12621-250
VWRU12621-236EA 174 EUR
VWRU12621-236 VWRU21570-127 VWRU12621-238 VWRU21570-404 VWRU12621-240 VWRU12621-250 241-0404 241-0405 241-0406
VWR® Talon®, Clamps, Three Pronged
Clamps General Purpose Clamps
Stainless steel or nickel-plated zinc. These clamps securely hold every type of glassware and apparatus.
  • Extension arms allow placement of apparatus at various distances from lab frames without compromising the integrity of the experiment
  • Stainless steel clamps: Exceptional chemical resistance, clamps are electropolished and autoclavable
  • Nickel-plated zinc clamps: High tensile strength
  • Supplied with non-slip vinyl sleeves, and fiberglass covers for temperatures above 100 °C
Long, seamless tubing attaches clamp head securely and offers easy positioningMore Product Information