Control Standard Endotoxin (CSE)
Supplier: FUJIFILM WAKO CHEMICALS
CSE4037-5006
WAKFCSE4037-5006EA 126 EUR
WAKFCSE4037-5006
Control Standard Endotoxin (CSE)
Microbiology Test Systems Endotoxin Tests
The Control Standard Endotoxin (CSE) is a lyophilised product composed of a 500 ng vial of endotoxin, purified from E. coli UKT-B strain that serves as an accurate and dependable standard for endotoxin testing.
  • Endotoxin derived from E. coli UKT-B
  • Can be used to prepare controls and standard curves
  • Potency of CSE supplied to match each lot of LAL
  • Reconstituted CSE can be stored at 2 to 10 °C for 1 month
Order Now
SPECIFICATIONS
Environmentally Preferable