Hydrobromic acid 47%

Supplier: Wako
Danger

Synonyms: Hydrogen bromide

639-04655
WAKO639-04655EA 578 EUR
WAKO639-04655
Hydrobromic acid 47%
Hydrogen bromide
Formula: HBr
MW: 80.91 g/mol
Boiling Pt: -66.7
Melting Pt: -86
Density: 1.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8 II
Merck Index: 13,04799

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR