Cap for BOD bottle, WHEATON®
Supplier: DWK Life Sciences
227723
WHEA227723EA 116 EUR
WHEA227723
Cap for BOD bottle, WHEATON®
Closures Bottle Closures
This polyethylene cap, complete with a closed cell foam insert, snaps firmly over the flared mouth of Wheaton BOD bottles.
  • Polyethylene cap prevents evaporation
  • Includes foam insert
This closure prevents the evaporation of the water seal during the five-day BOD incubation period. The foam insert fits into the cap and exerts pressure on the bottle stopper to hold it securely in place.