Wright's stain modified, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Danger

Synonyms: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue

WS16-500ML
WS16-500MLEA 111 EUR
WS16-500ML
Wright's stain modified, Sigma-Aldrich®
Wright's stain

Popular hematology stain used for differentially staining the cellular elements of blood. For dip, rack, and batch staining techniques.

Storage Temperature: Ambient
MDL Number: MFCD00082143
CAS Number: 68988-92-1
ADR: 3 III
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR