Hazardous Waste Filtration System
Supplier: Merck
YT30142HW YT3014257 YT4014259
YT30142HWEA 12310 EUR
YT30142HW YT3014257 YT4014259
Hazardous Waste Filtration System
Filters Membrane Filters
If your waste samples contain non-volatile or semi-volatile substances or metals, use this hazardous waste filtration system for saparation, extratction, and filtration. Developed in collaboration with the US Environmental Protection Agency (EPA), the system is designed specifically for the US EPA Extraction Procedure (EP) Toxicity Test and Toxicity Characteristic Leaching Procedure (TCLP). The device separates solid and liquid phases of waste samples for analysis.
  • Contains interior holder with PTFE coating to prevent heavy metal contamination
  • Allows easy
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Weight 12,3 kg