Universal Boxes, Aluminium
Supplier: ZARGES LEICHTBAU
40835 40810 40677
ZARG40835EA 343 EUR
ZARG40835 ZARG40810 ZARG40677
Universal Boxes, Aluminium
Boxes Transport Boxes
Aluminium, high-quality.
  • Lid with stainless steel hinge strip, two lid straps and foamed seal all round
  • Cast-aluminium stacking corners
  • High level of stability due to three fully-welded aluminium profile frames
  • Ergonomic heavy-duty sprung drop handles
  • Snap fasteners: can be secured with a plug lock, lead seals, protection against bursting open or a padlock (max. shackle thickness 6 mm)
Secure transportation, practical storage and protective packaging in one. The transported or stored item is securely packed and protected against external influences.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable