Calibration weights, OIML R111, class M2
Supplier: ZWIEBEL
E201.12.11
ZWIEE201.12.11EA 0 EUR
ZWIEE201.12.11
Calibration weights, OIML R111, class M2
Weights Calibration Weights
Weights made from brass. M2 weights are designed to calibrate masses of a lower accuracy class and be used with class II and III weighing instruments.
  • Ultra-compact and easy to handle
  • Weights are available as individual units or in sets (wooden box/base)